Order Entry
Denmark
Orders LinkContactUsLinkComponent
Test Tubes, with screw cap

Test Tubes, with screw cap

160 mm, 16 mm, GL 18
Catalog # HECH42782051
Test Tubes, with screw cap
Catalog # HECH42782051
Test Tubes, with screw cap
Test Tubes, with screw cap

160 mm, 16 mm, GL 18

Price
565,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents

AR®/soda glass, with PP screw cap.

  • Heavy wall
  • Round bottom
  • With thread GL18
Specifications
  • Ext.Ø:
    16 mm
  • Thread:
    GL 18
  • Length:
    160 mm
  • Seq:
    02