Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spoons, double-ended

Spoons, double-ended

210 mm, 25×35 mm
Catalog # 231-1219
Supplier:   HAMMACHER KARL
Spoons, double-ended
Catalog # 231-1219
Supplier:  HAMMACHER KARL
Spoons, double-ended
Spoons, double-ended

210 mm, 25×35 mm

Price
560,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Double-ended spoons made from WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).

Specifications
  • Length:
    210 mm
  • Seq:
    03
  • Spoon size:
    25×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.