Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spoons, double-ended

Spoons, double-ended

180 mm, 25×35 mm
  HAMMHWN112-18
Spoons, double-ended
  HAMMHWN112-18
Spoons, double-ended
Spoons, double-ended

180 mm, 25×35 mm

 
558,00 kr
   
1 * 1 Items
 

Double-ended spoons made from WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).

 
  • Length:
    180 mm
  • Seq:
    02
  • Spoon size:
    25×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.