Order Entry
Denmark
Orders LinkContactUsLinkComponent
Forceps, straight
Forceps, straight
Straight, fine tips, 90 mm
Catalog # HAMMHWC102-09
Supplier:   HAMMACHER KARL
Forceps, straight
Catalog # HAMMHWC102-09
Supplier:  HAMMACHER KARL
Forceps, straight
Forceps, straight
Straight, fine tips, 90 mm
Price
450,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
WIRONIT steel.
Specifications
  • Type:
    Straight, fine tips
  • Length:
    90 mm