Order Entry
Denmark
Orders LinkContactUsLinkComponent
Forceps, teethed
120 mm
  232-0067
Forceps, teethed
  232-0067
Forceps, teethed
Forceps, teethed
120 mm
697,00 kr
1 * 1 Items
WIRONIT steel (18/12 stainless steel with 18% chrome and 10 - 12 % nickel)
  • Teethed, straight
  • Resistant to corrosion
  • Anti-magnetic
  • Length:
    120 mm