Order Entry
Denmark
Orders LinkContactUsLinkComponent
Microscopic forceps

Microscopic forceps

130 mm
Catalog # 232-2154
Supplier:   HAMMACHER KARL
Microscopic forceps
Catalog # 232-2154
Supplier:  HAMMACHER KARL
Microscopic forceps
Microscopic forceps

130 mm

Price
583,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).

  • Delicate
  • Resistant to corrosion
  • Anti-magnetic
Specifications
  • Length:
    130 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.