Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thumb forceps, standard
Thumb forceps, standard
Straight, serrated, blunt, 180 mm
Catalog # 232-1927
Supplier:   HAMMACHER KARL
Thumb forceps, standard
Catalog # 232-1927
Supplier:  HAMMACHER KARL
Thumb forceps, standard
Thumb forceps, standard
Straight, serrated, blunt, 180 mm
Price
601,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Standard forceps made from Wironit steel.
Specifications
  • Type:
    Straight, serrated, blunt
  • Length:
    180 mm
  • Seq:
    05