Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thumb forceps, standard
Thumb forceps, standard
Straight, serrated, blunt, 145 mm
Catalog # HAMML-HWC052-14
Supplier:   HAMMACHER KARL
Thumb forceps, standard
Catalog # HAMML-HWC052-14
Supplier:  HAMMACHER KARL
Thumb forceps, standard
Thumb forceps, standard
Straight, serrated, blunt, 145 mm
Price
542,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Standard forceps made from Wironit steel.
Specifications
  • Type:
    Straight, serrated, blunt
  • Length:
    145 mm
  • Seq:
    03
  • Cat. No.:
    HAMML-HWC052-14
Anbefalinger