Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dissecting Forceps, Micro
Dissecting Forceps, Micro
Straight, with fine sharp tips, cross action, 100 mm
Catalog # HAMMHSO012-10
Supplier:   HAMMACHER KARL
Dissecting Forceps, Micro
Catalog # HAMMHSO012-10
Supplier:  HAMMACHER KARL
Dissecting Forceps, Micro
Dissecting Forceps, Micro
Straight, with fine sharp tips, cross action, 100 mm
Price
906,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Extra fine micro forceps made from stainless steel.
Specifications
  • Type:
    Straight, with fine sharp tips, cross action
  • Length:
    100 mm
  • Seq:
    03