Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dissecting Forceps, Micro
Dissecting Forceps, Micro
Curved, with fine sharp tips, 100 mm
Catalog # HAMMHSO009-10
Supplier:   HAMMACHER KARL
Dissecting Forceps, Micro
Catalog # HAMMHSO009-10
Supplier:  HAMMACHER KARL
Dissecting Forceps, Micro
Dissecting Forceps, Micro
Curved, with fine sharp tips, 100 mm
Price
851,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Extra fine micro forceps made from stainless steel.
Specifications
  • Type:
    Curved, with fine sharp tips
  • Length:
    100 mm
  • Seq:
    01