Serrefines
Serrefine, Straight, 35 mm
Katalog # 221-0051
Leverandør: HAMMACHER KARL
Gå til:
Product Details & Documents
These small forceps for clamping a blood vessel are made from stainless steel.
Specifikationer
- Type:Straight
- Description:Serrefine
- Length:35 mm
- Seq:03