Order Entry
Denmark
Orders LinkContactUsLinkComponent
Micro forceps
Micro forceps
No. 7, curved, very fine tips, 115 mm
Catalog # HAMMHSC603-11
Supplier:   HAMMACHER KARL
Micro forceps
Catalog # HAMMHSC603-11
Supplier:  HAMMACHER KARL
Micro forceps
Micro forceps
No. 7, curved, very fine tips, 115 mm
Price
739,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    No. 7, curved, very fine tips
  • Length:
    115 mm
  • Seq:
    04