Order Entry
Denmark
Orders LinkContactUsLinkComponent
Preparation forceps
Preparation forceps
Straight, fine, 115 mm
Catalog # HAMMHSC186-11
Supplier:   HAMMACHER KARL
Preparation forceps
Catalog # HAMMHSC186-11
Supplier:  HAMMACHER KARL
Preparation forceps
Preparation forceps
Straight, fine, 115 mm
Price
336,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Straight, fine
  • Length:
    115 mm
  • Seq:
    05