Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dressing forceps, standard
Standard, Straight, blunt, 130 mm
Catalog # 232-2179
Supplier:   HAMMACHER KARL
Dressing forceps, standard
Catalog # 232-2179
Supplier:  HAMMACHER KARL
Dressing forceps, standard
Dressing forceps, standard
Standard, Straight, blunt, 130 mm
Price
294,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Straight, blunt
  • Description:
    Standard
  • Length:
    130 mm
  • Seq:
    03