Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spoon spatulas

Spoon spatulas

170 mm, White
Catalog # 231-1344
Spoon spatulas
Catalog # 231-1344
Spoon spatulas
Spoon spatulas

170 mm, White

Price
224,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Porcelain, glazed white.

  • Broad shape
  • Excellent chemical resistance
  • Heat resistant up to 1000 °C
  • Designed for rough handling
Specifications
  • Colour:
    White
  • Length:
    170 mm