Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spoon spatulas

Spoon spatulas

125 mm, White
Catalog # 231-1342
Spoon spatulas
Catalog # 231-1342
Spoon spatulas
Spoon spatulas

125 mm, White

Price
203,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Porcelain, glazed white.

  • Broad shape
  • Excellent chemical resistance
  • Heat resistant up to 1000 °C
  • Designed for rough handling
Specifications
  • Colour:
    White
  • Length:
    125 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.