Order Entry
Denmark
Orders LinkContactUsLinkComponent
Powder pillows, iron
Powder pillows, iron
Iron (FerroVer®) reagent, 0,02 - 3,00 mg/l, 1,10-Phenanthroline, 25 ml, PP, 100
Catalog # HACH85499
Supplier:   Hach
Powder pillows, iron
Catalog # HACH85499
Supplier:  Hach
CAS Number:  
Powder pillows, iron
Powder pillows, iron
Iron (FerroVer®) reagent, 0,02 - 3,00 mg/l, 1,10-Phenanthroline, 25 ml, PP, 100
CAS Number:  
Price
450,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
Specifications
Specifications
  • Chemical method:
    1,10-Phenanthroline
  • Pk:
    100
  • Description:
    Iron (FerroVer®) reagent
  • Sample size:
    25 ml
  • Type:
    PP
  • Range:
    0,02 - 3,00 mg/l
Specifications