Order Entry
Denmark
Orders LinkContactUsLinkComponent
Redox standards

Redox standards

Standard redox solution, 300 mV at 25 °C, 500 mL
Catalog # REGERS300
Supplier:   REAGECON
Redox standards
Catalog # REGERS300
Supplier:  REAGECON
CAS Number:  
Redox standards

Standard redox solution, 300 mV at 25 °C, 500 mL

CAS Number:  
Price
939,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

The potentials quoted are of platinum electrode with reference to Ag/AgCl (3 M KCl).

Documents

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Standard redox solution, 300 mV at 25 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.