Order Entry
Denmark
Orders LinkContactUsLinkComponent
ISE standard solutions

ISE standard solutions

Standard solution sodium ions (ES/Na), 1 L
Catalog # 662-0380
Supplier:   WTW
ISE standard solutions
Catalog # 662-0380
Supplier:  WTW
CAS Number:  
ISE standard solutions
ISE standard solutions

Standard solution sodium ions (ES/Na), 1 L

CAS Number:  
Price
1.670,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

These standard solutions for calibration contain 10 g of the ion to be measured per 10 liters.

Documents

Specifikationer

Specifikationer

  • Pk:
    1 L
  • Description:
    Standard solution sodium ions (ES/Na)

Specifikationer