Order Entry
Denmark
Orders LinkContactUsLinkComponent
Vial racks, WHEATON®

Vial racks, WHEATON®

12,5 mm, 50, 190×100×22 mm
Catalog # 212-6192
Supplier:   DWK Life Sciences
Vial racks, WHEATON®
Catalog # 212-6192
Supplier:  DWK Life Sciences
Vial racks, WHEATON®
Vial racks, WHEATON®

12,5 mm, 50, 190×100×22 mm

Price
2.670,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents

These polypropylene racks accommodate 36 or 50 headspace auto-sampler vials. Each well has an opening in the bottom to facilitate drainage.

  • Autoclavable and stackable
  • Chemically resistant
  • Include position identifiers
Specifications
  • Hole Ø:
    12,5 mm
  • No. of Holes:
    50
  • W×D×H:
    190×100×22 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.