Order Entry
Denmark
Orders LinkContactUsLinkComponent
Trypsinising flasks, with screw caps, WHEATON®
Trypsinising flasks, with screw caps, WHEATON®
150 ml, Top screw cap 38-430
Catalog # WHEA355394
Supplier:   DWK Life Sciences
Trypsinising flasks, with screw caps, WHEATON®
Catalog # WHEA355394
Supplier:  DWK Life Sciences
Trypsinising flasks, with screw caps, WHEATON®
Trypsinising flasks, with screw caps, WHEATON®
150 ml, Top screw cap 38-430
Price
1.060,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
Certifications: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
Specifications
  • Capacity:
    150 ml
  • Description:
    Top screw cap 38-430
Anbefalinger