Order Entry
Denmark
Orders LinkContactUsLinkComponent
Conductivity Standards

Conductivity Standards

Conductivity reference standard, 5000 µS/cm ± 1%
Catalog # REGECRMCSKC5M
New Product
Supplier:   REAGECON
Conductivity Standards
Catalog # REGECRMCSKC5M
Supplier:  REAGECON
New Product
CAS Number:  
Conductivity Standards

Conductivity reference standard, 5000 µS/cm ± 1%

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents

To enable analysts to achieve the correct result when measuring conductivity and to prove the correctness of that result; Reagecon has developed a high quality range of Conductivity Standards; which can be used for calibration of conductivity sensors; quality control; method validation and instrument qualification.

  • Reagecon Conductivity Standards are aqueous based; thereby eliminating any errors attributable to matrix mismatch

    Directly traceable to NIST Standard Reference Materials; meets the specification requirements of all the major Pharmacopoeias

    The low temperature coefficient of variation; reduces measurement errors and enables non-temperature controlled use and is suitable for field work

    High accuracy ±1% at 25 °C; enables the Standards to be used as calibrators and/or controls; in accordance with the most exacting industrial requirements

    Manufactured to exacting specifications with an extended shelf life and stability; even after opening the bottle thereby eliminating the requirement to open a fresh bottle of Standard every time it is used

Tested at 25 °C

Used with conductivity meters

Packaging: HDPE Bottle GL 45

Physical Form: Liquid

Specifications

Specifications

  • Description:
    Conductivity reference standard
  • Conductivity:
    5000 µS/cm ± 1%

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.