Order Entry
Denmark
Orders LinkContactUsLinkComponent
TexWrite® Spiral Cleanroom Notebooks

TexWrite® Spiral Cleanroom Notebooks

Notebook, spiral, White, TX5741, Ruled, 140×216 mm, 100
Catalog # TEXWTX5741
New Product
Supplier:   Texwipe
TexWrite® Spiral Cleanroom Notebooks
Catalog # TEXWTX5741
Supplier:  Texwipe
New Product
TexWrite® Spiral Cleanroom Notebooks
TexWrite® Spiral Cleanroom Notebooks

Notebook, spiral, White, TX5741, Ruled, 140×216 mm, 100

Price
2.120,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

TexWrite® cleanroom spiral-bound notebook, college ruled. An economical choice for note taking and data recording in critical environments.

  • Printed with low sodium, IPA resistant inks
  • Lie flat and pages rotate freely for ease of use

Format: large (279×216 mm), medium (216×140 mm), small (127×76 mm).

Packaging: Bag-Within-A-Bag®.

Specifications
  • Paper size:
    140×216 mm
  • Type:
    Ruled
  • Colour:
    White
  • Description:
    Notebook, spiral
  • Seq:
    4
  • No. of pages:
    100
  • Model:
    TX5741
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.