Order Entry
Denmark
Orders LinkContactUsLinkComponent
Phenol Red indicator

Phenol Red indicator

Bottle
Catalog # APOSOR1014625-100G
Irriterende
New Product
Supplier:   Apollo Scientific
Phenol Red indicator
Catalog # APOSOR1014625-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  143-74-8
Phenol Red indicator

Bottle

CAS Number:  143-74-8
Price
231,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

Synonymer:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS nummer:143-74-8
  • Formel:C₁₉H₁₄O₅S
  • MDL nummer:MFCD00003552
  • Molecular Weight:354,38 g/mol
  • EINECS:205-609-7
Specifikationer

Specifikationer

  • Pack type:
    Bottle

Specifikationer