Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-Sorbitol ≥95%

D-Sorbitol ≥95%

Bottle
Catalog # APOSBUP14992-500MG
New Product
Supplier:   Apollo Scientific
D-Sorbitol ≥95%
Catalog # APOSBUP14992-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  50-70-4
D-Sorbitol ≥95%

Bottle

CAS Number:  50-70-4
Price
kr538.00
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  • CAS Number:50-70-4
  • Formula:C₆H₁₄O₆
  • Molecular Weight:182,17 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications