Murexide indicator
Bottle
Catalog # APOSOR2362-25G
New Product
Supplier: Apollo Scientific
Murexide indicator
Catalog # APOSOR2362-25G
Supplier: Apollo Scientific
New Product
CAS Number: 3051-09-0
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS Number:3051-09-0
- Formula:C₈H₅N₅O₆·H₃N
- MDL Number:MFCD00012777
- Molecular Weight:284,18572 g/mol
- EINECS:221-266-6
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



