Biotin hydrazide
Bottle
APOSBIB108-1G
New Product
Carbohydrate reactive. Microscopy studies on membranesRef: O'Shannessy,D.J.et al. (1984) Immun.Letters 8,273-277
: 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide
- :66640-86-6
- :C₁₀H₁₈N₄O₂S
- :MFCD00078532
- :258.34 g/mol
- :240…244 °C
- :Refrigerator
- Pack type:Bottle