Order Entry
Denmark
Orders LinkContactUsLinkComponent
Murexide indicator

Murexide indicator

Bottle
Catalog # APOSOR2362-500G
Irritant
New Product
Supplier:   Apollo Scientific
Murexide indicator
Catalog # APOSOR2362-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  3051-09-0
Murexide indicator

Bottle

CAS Number:  3051-09-0
Price
1.200,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
  • CAS Number:3051-09-0
  • Formula:C₈H₅N₅O₆·H₃N
  • MDL Number:MFCD00012777
  • Molecular Weight:284,18572 g/mol
  • EINECS:221-266-6
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.