Order Entry
Denmark
Orders LinkContactUsLinkComponent
Biotin hydrazide

Biotin hydrazide

Bottle
Catalog # APOSBIB108-25G
Irritant
New Product
Supplier:   Apollo Scientific
Biotin hydrazide
Catalog # APOSBIB108-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  66640-86-6
Biotin hydrazide

Bottle

CAS Number:  66640-86-6
Price
16.390,00 kr
Log in for availability
1 * 25 g
Product Details & Documents

Carbohydrate reactive. Microscopy studies on membranesRef: O'Shannessy,D.J.et al. (1984) Immun.Letters 8,273-277

Synonyms:  5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanehydrazide
  • CAS Number:66640-86-6
  • Formula:C₁₀H₁₈N₄O₂S
  • MDL Number:MFCD00078532
  • Molecular Weight:258.34 g/mol
  • Melting Pt:240…244 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle

Specifications