DIPSO ≥98%
Bottle
Catalog # APOSBIB1158-5G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Biological buffer - useful range 7.0 - 8.2.
US Patent No. 4,169,950 (To Research Organics)
Synonyms: 3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
- CAS Number:68399-80-4
- Formula:C₇H₁₇NO₆S
- MDL Number:MFCD00038353
- Molecular Weight:261.29 g/mol
- Melting Pt:189…192 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSBIB1158-5G
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



