Order Entry
Denmark
Orders LinkContactUsLinkComponent
DIPSO ≥98%

DIPSO ≥98%

Bottle
Catalog # APOSBIB1158-2.5KG
New Product
Supplier:   Apollo Scientific
DIPSO ≥98%
Catalog # APOSBIB1158-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  68399-80-4
DIPSO ≥98%

Bottle

CAS Number:  68399-80-4
Price
4.560,00 kr
Log in for availability
1 * 2,5 kg
Product Details & Documents

Biological buffer - useful range 7.0 - 8.2.

US Patent No. 4,169,950 (To Research Organics)

Synonyms:  3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
  • CAS Number:68399-80-4
  • Formula:C₇H₁₇NO₆S
  • MDL Number:MFCD00038353
  • Molecular Weight:261.29 g/mol
  • Melting Pt:189…192 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.