Order Entry
Denmark
Orders LinkContactUsLinkComponent
Caffeic acid

Caffeic acid

Bottle
Catalog # APOSOR1165-5G
Irritant
Health
New Product
Supplier:   Apollo Scientific
Caffeic acid
Catalog # APOSOR1165-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  331-39-5
Caffeic acid

Bottle

CAS Number:  331-39-5
Price
165,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
  • CAS Number:331-39-5
  • Formula:C₉H₈O₄
  • MDL Number:MFCD00004392
  • Molecular Weight:180.16 g/mol
  • Melting Pt:194…198 °C
  • Storage Temperature:Ambient
  • EINECS:206-361-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.