Order Entry
Denmark
Orders LinkContactUsLinkComponent
1-Hydroxyurea 98%

1-Hydroxyurea 98%

Bottle
Catalog # APOSOR9108-500G
Health
New Product
Supplier:   Apollo Scientific
1-Hydroxyurea 98%
Catalog # APOSOR9108-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  127-07-1
1-Hydroxyurea 98%

Bottle

CAS Number:  127-07-1
Price
396,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • Store at 2 to 8 °C
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76,05466 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.