Order Entry
Denmark
Orders LinkContactUsLinkComponent
1-Hydroxyurea 98%

1-Hydroxyurea 98%

Bottle
Catalog # APOSOR9108-100G
Health
New Product
Supplier:   Apollo Scientific
1-Hydroxyurea 98%
Catalog # APOSOR9108-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  127-07-1
1-Hydroxyurea 98%

Bottle

CAS Number:  127-07-1
Price
187,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • Store at 2 to 8 °C
Synonyms:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS Number:127-07-1
  • Formula:CH₄N₂O₂
  • MDL Number:MFCD00007943
  • Molecular Weight:76,05466 g/mol
  • Storage Temperature:Refrigerator
  • EINECS:204-821-7
Specifications

Specifications

  • Pack type:
    Bottle

Specifications