Order Entry
Denmark
Orders LinkContactUsLinkComponent
Thymolphthalein indicator

Thymolphthalein indicator

Bottle
Catalog # APOSOR1015241-5G
New Product
Supplier:   Apollo Scientific
Thymolphthalein indicator
Catalog # APOSOR1015241-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  125-20-2
Thymolphthalein indicator

Bottle

CAS Number:  125-20-2
Price
165,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430,54 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications