Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cholesterol 95%

Cholesterol 95%

Bottle
Catalog # APOSOR1030208-100G
Irritant
New Product
Supplier:   Apollo Scientific
Cholesterol 95%
Catalog # APOSOR1030208-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  57-88-5
Cholesterol 95%

Bottle

CAS Number:  57-88-5
Price
286,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
  • CAS Number:57-88-5
  • Formula:C₂₇H₄₆O
  • MDL Number:MFCD00003646
  • Molecular Weight:386,55 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.