Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cyclic AMP ≥98%

Cyclic AMP ≥98%

Bottle
Catalog # APOSBUP17291-1G
New Product
Supplier:   Apollo Scientific
Cyclic AMP ≥98%
Catalog # APOSBUP17291-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  60-92-4
Cyclic AMP ≥98%

Bottle

CAS Number:  60-92-4
Price
802,00 kr
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  3',5'-cAMP, 3′,5′-Cyclic AMP, Adenosine 3′,5′-cyclophosphate, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide
  • CAS Number:60-92-4
  • Formula:C₁₀H₁₂N₅O₆P
  • Molecular Weight:329,21 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.