Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cyclic AMP ≥98%

Cyclic AMP ≥98%

Bottle
Catalog # APOSBUP17291-500MG
New Product
Supplier:   Apollo Scientific
Cyclic AMP ≥98%
Catalog # APOSBUP17291-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  60-92-4
Cyclic AMP ≥98%

Bottle

CAS Number:  60-92-4
Price
472,00 kr
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  3',5'-cAMP, 3′,5′-Cyclic AMP, Adenosine 3′,5′-cyclophosphate, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide
  • CAS Number:60-92-4
  • Formula:C₁₀H₁₂N₅O₆P
  • Molecular Weight:329,21 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications