Order Entry
Denmark
Orders LinkContactUsLinkComponent
Aluminon

Aluminon

Bottle
Catalog # APOSOR2429-500G
Irritant
New Product
Supplier:   Apollo Scientific
Aluminon
Catalog # APOSOR2429-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  569-58-4
Aluminon

Bottle

CAS Number:  569-58-4
Price
615,00 kr
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
  • CAS Number:569-58-4
  • Formula:C₂₂H₂₃N₃O₉
  • MDL Number:MFCD00040925
  • Molecular Weight:473.44 g/mol
  • Melting Pt:220…225 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:209-319-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.