Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium L(+)-ascorbate 98%

Sodium L(+)-ascorbate 98%

Bottle
Catalog # APOSOR922808-100G
New Product
Supplier:   Apollo Scientific
Sodium L(+)-ascorbate 98%
Catalog # APOSOR922808-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  134-03-2
Sodium L(+)-ascorbate 98%

Bottle

CAS Number:  134-03-2
Price
220,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
  • CAS Number:134-03-2
  • Formula:C₆H₇NaO₆
  • MDL Number:MFCD00082340
  • Molecular Weight:198.11 g/mol
  • Melting Pt:219…221 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.