Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

D-Mannitol ≥98%

Bottle
Catalog # APOSBUP18757-5G
New Product
Supplier:   Apollo Scientific
D-Mannitol ≥98%
Catalog # APOSBUP18757-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  69-65-8
D-Mannitol ≥98%

Bottle

CAS Number:  69-65-8
Price
538,00 kr
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:C₆H₁₄O₆
  • Molecular Weight:182,17 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications