Order Entry
Denmark
Orders LinkContactUsLinkComponent
Wright Stain 97%

Wright Stain 97%

Bottle
Catalog # APOSOR1027942-2.5K
New Product
Supplier:   Apollo Scientific
Wright Stain 97%
Catalog # APOSOR1027942-2.5K
Supplier:  Apollo Scientific
New Product
CAS Number:  68988-92-1
Wright Stain 97%

Bottle

CAS Number:  68988-92-1
Price
7.570,00 kr
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
  • CAS Number:68988-92-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.