Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Docusate sodium ≥98%

Bottle
  APOSBUP17054-500MG
New Product
Docusate sodium ≥98%
  APOSBUP17054-500MG
New Product
 :  577-11-7
Docusate sodium ≥98%

Bottle

 :  577-11-7
 
1.200,00 kr
   
1 * 500 mg
 

Usage: Biochemical/life science applications.

 :  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  •  :577-11-7
  •  :C₂₀H₃₇NaO₇S
  •  :444,56 g/mol
 

 

  • Pack type:
    Bottle