Order Entry
Denmark
Orders LinkContactUsLinkComponent
Docusate sodium ≥98%

Docusate sodium ≥98%

Bottle
Catalog # APOSBUP17054-200MG
New Product
Supplier:   Apollo Scientific
Docusate sodium ≥98%
Catalog # APOSBUP17054-200MG
Supplier:  Apollo Scientific
New Product
CAS Number:  577-11-7
Docusate sodium ≥98%

Bottle

CAS Number:  577-11-7
Price
725,00 kr
Log in for availability
1 * 200 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Dioctyl sulphosuccinate sodium salt, Dioctyl sulphosuccinate, sodium salt, Sodium dioctyl sulphosuccinate, Sodium 1,4-bis((2-ethylhexyl)oxy)-1,4-dioxobutane-2-sulphonate, bis(2-Ethylhexyl) sodium sulphosuccinate
  • CAS Number:577-11-7
  • Formula:C₂₀H₃₇NaO₇S
  • Molecular Weight:444,56 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.