Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzo[a]pyrene 99%

Benzo[a]pyrene 99%

Bottle
Catalog # APOSOR1025571-250M
New Product
Supplier:   Apollo Scientific
Benzo[a]pyrene 99%
Catalog # APOSOR1025571-250M
Supplier:  Apollo Scientific
New Product
CAS Number:  50-32-8
Benzo[a]pyrene 99%

Bottle

CAS Number:  50-32-8
Price
615,00 kr
Log in for availability
1 * 250 mg
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Benzo[def]chrysene, BaP
  • CAS Number:50-32-8
  • Formula:C₂₀H₁₂
  • Molecular Weight:252,31 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.