Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzo[a]pyrene 99%

Benzo[a]pyrene 99%

Bottle
Catalog # APOSOR1025571-100M
New Product
Supplier:   Apollo Scientific
Benzo[a]pyrene 99%
Catalog # APOSOR1025571-100M
Supplier:  Apollo Scientific
New Product
CAS Number:  50-32-8
Benzo[a]pyrene 99%

Bottle

CAS Number:  50-32-8
Price
308,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  Benzo[def]chrysene, BaP
  • CAS Number:50-32-8
  • Formula:C₂₀H₁₂
  • Molecular Weight:252,31 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications