Order Entry
Denmark
Orders LinkContactUsLinkComponent
Lead (II) Carbonate basic 99%

Lead (II) Carbonate basic 99%

Bottle
Catalog # APOSIN2213-25KG
New Product
Supplier:   Apollo Scientific
Lead (II) Carbonate basic 99%
Catalog # APOSIN2213-25KG
Supplier:  Apollo Scientific
New Product
CAS Number:  1319-46-6
Lead (II) Carbonate basic 99%

Bottle

CAS Number:  1319-46-6
Price
19.210,00 kr
Log in for availability
1 * 25 kg
Product Details & Documents

Usage: Inorganic salt or catalyst.

Synonyms:  Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate
  • CAS Number:1319-46-6
  • Formula:H₂O₂Pb·2CO₃·2Pb
  • MDL Number:MFCD00078155
  • Molecular Weight:775,63248 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications