Order Entry
Denmark
Orders LinkContactUsLinkComponent
Lead (II) Carbonate basic 99%

Lead (II) Carbonate basic 99%

Bottle
Catalog # APOSIN2213-2.5KG
New Product
Supplier:   Apollo Scientific
Lead (II) Carbonate basic 99%
Catalog # APOSIN2213-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  1319-46-6
Lead (II) Carbonate basic 99%

Bottle

CAS Number:  1319-46-6
Price
1.980,00 kr
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Inorganic salt or catalyst.

Synonyms:  Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate
  • CAS Number:1319-46-6
  • Formula:H₂O₂Pb·2CO₃·2Pb
  • MDL Number:MFCD00078155
  • Molecular Weight:775,63248 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.