Lead (II) Carbonate basic 99%
Bottle
Catalog # APOSIN2213-500G
New Product
Supplier: Apollo Scientific
Lead (II) Carbonate basic 99%
Catalog # APOSIN2213-500G
Supplier: Apollo Scientific
New Product
CAS Number: 1319-46-6
Jump to:
Product Details & Documents
Usage: Inorganic salt or catalyst.
Synonyms: Lead carbonat basic, White lead, Lead (II) carbonate hydroxide, Lead (II) hydroxide carbonate
- CAS Number:1319-46-6
- Formula:H₂O₂Pb·2CO₃·2Pb
- MDL Number:MFCD00078155
- Molecular Weight:775,63248 g/mol
Specifications
Specifications
- Pack type:Bottle



