Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-Ribose 99%
Bottle
  APOSBIF5012-100G
New Product
D-Ribose 99%
  APOSBIF5012-100G
New Product
 :  50-69-1
D-Ribose 99%
Bottle
 :  50-69-1
187,00 kr
1 * 100 g
Usage: Biochemical/ life science applications.
  • Store at 2 to 8 °C
  • Non-hazardous
  •  :50-69-1
  •  :C₅H₁₀O₅
  •  :MFCD00135453
  •  :150,1299 g/mol
  •  :Refrigerator
  •  :200-059-4
  • Pack type:
    Bottle